Assay Details
Target Gene Details
Entrez Gene ID: | 100130950 |
Gene Name: | uncharacterized LOC100130950 |
Gene Aliases: |
- |
Location: |
Chr.17:5192084-5235636 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100130950 | NR_034082.1 | |||
| AK021878.1 |
Target Gene Details
Entrez Gene ID: | 388325 |
Gene Name: | SLP adaptor and CSK interacting membrane protein |
Gene Aliases: |
C17orf87, UNQ5783 |
Location: |
Chr.17:5208920-5234860 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCIMP | NM_001271842.1 | NP_001258771.1 | ||
| NM_001319190.1 | NP_001306119.1 | |||
| NM_207103.3 | NP_996986.1 | |||
| XM_017024615.1 | XP_016880104.1 | |||
| AK057142.1 | ||||
| AK297035.1 | ||||
| AK310145.1 | ||||
| AY358809.1 | AAQ89169.1 | |||
| BC127925.1 | ||||
| BC130554.1 | ||||
| BC130556.1 | ||||
| BC144395.1 | ||||
| DQ778084.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
| dgv3089n100 | Chr.17:5187013 - 5327634 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map