Assay Details
Target Gene Details
Entrez Gene ID: | 131890 |
Gene Name: | G protein-coupled receptor kinase 7 |
Gene Aliases: |
GPRK7 |
Location: |
Chr.3:141763127-141817373 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRK7 | NM_139209.2 | 1 | 591 | NP_631948.1 |
| XM_017005717.1 | 3 | 3333 | XP_016861206.1 | |
| AF282269.1 | 1 | 591 | AAL33880.1 | |
| AF439409.1 | 1 | 644 | AAL48216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819618 | Chr.3:141778191 - 141779075 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map