Assay Details
Target Gene Details
Entrez Gene ID: | 41 |
Gene Name: | acid sensing ion channel subunit 1 |
Gene Aliases: |
ACCN2, ASIC, BNaC2 |
Location: |
Chr.12:50057637-50083622 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASIC1 | NM_001095.3 | NP_001086.2 | ||
| NM_020039.3 | NP_064423.2 | |||
| NR_046389.1 | ||||
| XM_011538350.1 | XP_011536652.1 | |||
| XM_011538351.2 | XP_011536653.1 | |||
| XM_011538352.1 | XP_011536654.1 | |||
| AK290010.1 | ||||
| BC133707.1 | ||||
| EU078959.1 | ||||
| U78180.1 | AAB48980.1 | |||
| U78181.1 | AAB48981.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv558837 | Chr.12:49955982 - 50073986 on Build GRCh38 | Loss |
|
| nsv558840 | Chr.12:50056193 - 50130181 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map