Assay Details
Target Gene Details
Entrez Gene ID: | 57568 |
Gene Name: | signal induced proliferation associated 1 like 2 |
Gene Aliases: |
SPAL2 |
Location: |
Chr.1:232397965-232630491 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIPA1L2 | NM_020808.4 | 21 | 6200 | NP_065859.3 |
| XM_005273211.2 | 23 | 6537 | XP_005273268.1 | |
| XM_005273212.4 | 23 | 6344 | XP_005273269.1 | |
| XM_005273213.4 | 22 | 6237 | XP_005273270.1 | |
| XM_011544243.2 | 24 | 6393 | XP_011542545.1 | |
| XM_017001896.1 | 23 | 6290 | XP_016857385.1 | |
| XM_017001897.1 | 22 | 6483 | XP_016857386.1 | |
| XM_017001898.1 | 22 | 6257 | XP_016857387.1 | |
| AB037810.1 | 21 | 5404 | BAA92627.1 | |
| AY168879.1 | 21 | 6160 | AAO12530.1 | |
| BC013119.1 | 12 | 2657 | ||
| BC150334.1 | 21 | 5415 | ||
| CB242802.1 | 1 | 413 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006763 | Chr.1:232341238 - 232458244 on Build GRCh38 | Gain |
|
| nsv549355 | Chr.1:232388426 - 232435636 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map