Assay Details
Target Gene Details
Entrez Gene ID: | 7227 |
Gene Name: | transcriptional repressor GATA binding 1 |
Gene Aliases: |
GC79, LGCR |
Location: |
Chr.8:115408496-115669633 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPS1 | NM_001282902.2 | 6 | 7905 | NP_001269831.1 |
| NM_001282903.2 | 7 | 8394 | NP_001269832.1 | |
| NM_014112.4 | 7 | 8408 | NP_054831.2 | |
| XM_005251049.2 | 6 | 8223 | XP_005251106.1 | |
| XM_006716625.1 | 7 | 8345 | XP_006716688.1 | |
| XM_011517264.1 | 8 | 8478 | XP_011515566.1 | |
| XM_011517266.2 | 7 | 8213 | XP_011515568.1 | |
| XM_011517267.1 | 7 | 8331 | XP_011515569.1 | |
| XM_011517268.1 | 7 | 8320 | XP_011515570.1 | |
| XM_017013801.1 | 6 | 12007 | XP_016869290.1 | |
| AF183810.1 | 7 | 8402 | AAF23614.1 | |
| AK021511.1 | 1 | 1415 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428205 | Chr.8:115329343 - 115510682 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map