Assay Details
Target Gene Details
Entrez Gene ID: | 5264 |
Gene Name: | phytanoyl-CoA 2-hydroxylase |
Gene Aliases: |
LN1, LNAP1, PAHX, PHYH1, RD |
Location: |
Chr.10:13277796-13300130 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHYH | NM_001037537.1 | 8 | 1547 | NP_001032626.1 |
| NM_001323080.1 | 9 | 1606 | NP_001310009.1 | |
| NM_001323082.1 | 9 | 1448 | NP_001310011.1 | |
| NM_001323083.1 | 7 | 1178 | NP_001310012.1 | |
| NM_001323084.1 | 8 | 1553 | NP_001310013.1 | |
| NM_006214.3 | 9 | 1442 | NP_006205.1 | |
| AF023462.1 | 9 | 1381 | AAB81834.1 | |
| AF112977.1 | 9 | 1413 | AAD20602.1 | |
| BC029512.1 | 9 | 1402 | AAH29512.1 | |
| BM351828.1 | 1 | 183 |
More Information
Set Membership: |
|

Genomic Map