Assay Details
Target Gene Details
Entrez Gene ID: | 25915 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 3 |
Gene Aliases: |
2P1, C3orf60, E3-3 |
Location: |
Chr.3:49020475-49023495 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFAF3 | NM_199069.1 | 1 | 364 | NP_951032.1 |
| NM_199070.1 | NP_951033.1 | |||
| NM_199073.1 | NP_951047.1 | |||
| NM_199074.1 | NP_951056.1 | |||
| AL049955.1 | CAB43224.2 | |||
| BC111004.1 | 1 | 994 | ||
| BQ652614.1 | ||||
| CD358942.1 |
More Information
Set Membership: |
|

Genomic Map