Assay Details
Target Gene Details
Entrez Gene ID: | 10225 |
Gene Name: | CD96 molecule |
Gene Aliases: |
TACTILE |
Location: |
Chr.3:111542079-111665996 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD96 | NM_001318889.1 | 2 | 317 | NP_001305818.1 |
| NM_005816.4 | 2 | 317 | NP_005807.1 | |
| NM_198196.2 | 2 | 317 | NP_937839.1 | |
| NR_134917.1 | 2 | 317 | ||
| XM_005247063.3 | 2 | 278 | XP_005247120.1 | |
| XM_006713469.3 | 2 | 278 | XP_006713532.1 | |
| XM_006713470.3 | 2 | 278 | XP_006713533.1 | |
| XM_017005521.1 | 2 | 278 | XP_016861010.1 | |
| XM_017005522.1 | 2 | 278 | XP_016861011.1 | |
| AK290719.1 | 2 | 203 | ||
| AK315237.1 | 2 | 199 | ||
| AL833681.1 | 2 | 164 | CAI46155.1 | |
| AW341041.1 | 2 | 297 | ||
| BC020749.1 | 2 | 248 | AAH20749.1 | |
| BM561433.1 | 2 | 317 | ||
| DB130968.1 | 2 | 302 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591275 | Chr.3:110918624 - 111782250 on Build GRCh38 | Gain |
|
| nsv3946 | Chr.3:111517188 - 111564191 on Build GRCh38 | Deletion |
|
| nsv997756 | Chr.3:111507211 - 111657738 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map