Assay Details
Target Gene Details
Entrez Gene ID: | 8847 |
Gene Name: | deleted in lymphocytic leukemia 2 (non-protein coding) |
Gene Aliases: |
1B4, BCMSUN, DLB2, LEU2, LINC00022, MIR15AHG, NCRNA00022, RFP2OS, TRIM13OS |
Location: |
Chr.13:49982552-50125541 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLEU2 | NR_002612.1 | |||
| AY455757.1 |
Target Gene Details
Entrez Gene ID: | 10206 |
Gene Name: | tripartite motif containing 13 |
Gene Aliases: |
CAR, DLEU5, LEU5, RFP2, RNF77 |
Location: |
Chr.13:49997007-50018467 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM13 | NM_001007278.2 | NP_001007279.1 | ||
| NM_005798.4 | NP_005789.2 | |||
| NM_052811.3 | NP_434698.1 | |||
| NM_213590.2 | NP_998755.1 | |||
| AF220127.1 | AAG53500.1 | |||
| AF241850.1 | AAF91315.1 | |||
| AJ224819.1 | CAA12136.1 | |||
| AK314496.1 | ||||
| AL360196.1 | ||||
| AY191002.1 | AAO38979.1 | |||
| AY455758.1 | AAR31110.1 | |||
| AY764035.1 | AAV51406.1 | |||
| BC003579.1 | AAH03579.1 | |||
| BC063407.1 | AAH63407.1 | |||
| BI668033.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv561618 | Chr.13:49907577 - 51521449 on Build GRCh38 | Loss |
|
| nsv1035151 | Chr.13:49939399 - 50593714 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map