Assay Details
Target Gene Details
Entrez Gene ID: | 93210 |
Gene Name: | post-GPI attachment to proteins 3 |
Gene Aliases: |
AGLA546, CAB2, PERLD1, PP1498, hCOS16 |
Location: |
Chr.17:39671122-39688070 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGAP3 | NM_001291726.1 | 7 | 1763 | NP_001278655.1 |
| NM_001291728.1 | 7 | 1853 | NP_001278657.1 | |
| NM_001291730.1 | 6 | 1574 | NP_001278659.1 | |
| NM_001291732.1 | 5 | 1511 | NP_001278661.1 | |
| NM_001291733.1 | 4 | 1449 | NP_001278662.1 | |
| NM_033419.4 | 8 | 1916 | NP_219487.3 | |
| XM_011525480.2 | 7 | 1698 | XP_011523782.1 | |
| XM_011525481.2 | 9 | 2045 | XP_011523783.1 | |
| AB088396.1 | 8 | 1868 | BAC55580.1 | |
| AF217980.1 | 5 | 1311 | AAG17223.1 | |
| AK057911.1 | 8 | 2224 | ||
| AK075473.1 | 1 | 91 | ||
| AK075474.1 | 7 | 1750 | BAC11642.1 | |
| AK289612.1 | 4 | 1436 | ||
| AL833198.1 | 1 | 614 | ||
| AY358437.1 | 8 | 1892 | AAQ88803.1 | |
| BC010652.1 | 8 | 1877 | AAH10652.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457738 | Chr.17:39613752 - 39674782 on Build GRCh38 | Loss |
|
| nsv516757 | Chr.17:39625596 - 39678288 on Build GRCh38 | Loss |
|
| nsv528093 | Chr.17:39657827 - 39709752 on Build GRCh38 | Loss |
|
| dgv40n68 | Chr.17:39536627 - 39726611 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map