Assay Details
Target Gene Details
Entrez Gene ID: | 617 |
Gene Name: | BCS1 homolog, ubiquinol-cytochrome c reductase complex chaperone |
Gene Aliases: |
BCS, BCS1, BJS, FLNMS, GRACILE, Hs.6719, MC3DN1, PTD, h-BCS, h-BCS1 |
Location: |
Chr.2:218659656-218663443 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCS1L | NM_001079866.1 | NP_001073335.1 | ||
| NM_001257342.1 | NP_001244271.1 | |||
| NM_001257343.1 | NP_001244272.1 | |||
| NM_001257344.1 | NP_001244273.1 | |||
| NM_001318836.1 | NP_001305765.1 | |||
| NM_001320717.1 | NP_001307646.1 | |||
| NM_004328.4 | NP_004319.1 | |||
| XM_005246749.3 | XP_005246806.1 | |||
| XM_017004633.1 | XP_016860122.1 | |||
| BP215082.1 | ||||
| DA540458.1 | ||||
| DA613959.1 |
Target Gene Details
Entrez Gene ID: | 7701 |
Gene Name: | zinc finger protein 142 |
Gene Aliases: |
pHZ-49 |
Location: |
Chr.2:218637592-218659632 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF142 | NR_073599.1 | |||
| XM_006712730.3 | XP_006712793.1 | |||
| XM_011511784.2 | XP_011510086.1 | |||
| XM_011511785.2 | XP_011510087.1 | |||
| XM_011511786.2 | XP_011510088.1 | |||
| XM_011511787.2 | XP_011510089.1 | |||
| XM_011511788.2 | XP_011510090.1 | |||
| XM_011511789.2 | XP_011510091.1 |
More Information
Set Membership: |
|

Genomic Map