Assay Details
Target Gene Details
Entrez Gene ID: | 842 |
Gene Name: | caspase 9 |
Gene Aliases: |
APAF-3, APAF3, ICE-LAP6, MCH6, PPP1R56 |
Location: |
Chr.1:15491401-15524912 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASP9 | NM_001229.4 | NP_001220.2 | ||
| NM_001278054.1 | NP_001264983.1 | |||
| NM_032996.3 | NP_127463.2 | |||
| NR_102732.1 | ||||
| NR_102733.1 | ||||
| XM_005246014.2 | XP_005246071.1 | |||
| AB015653.1 | BAA78780.1 | |||
| AB020979.1 | BAA87905.1 | |||
| AB209147.1 | 8 | 2308 | BAD92384.1 | |
| AF093130.1 | AAD12248.1 | |||
| AF110376.1 | AAD13615.1 | |||
| AJ781267.1 | CAH03131.1 | |||
| AK292080.1 | ||||
| AK292111.1 | ||||
| AK303743.1 | ||||
| AY732490.2 | AAV33129.1 | |||
| BC002452.2 | AAH02452.1 | |||
| BC006463.1 | AAH06463.1 | |||
| BT006911.1 | AAP35557.1 | |||
| U56390.1 | AAC50640.1 | |||
| U60521.1 | AAC50776.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833536 | Chr.1:15323505 - 15528830 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map