Assay Details
Target Gene Details
Entrez Gene ID: | 6520 |
Gene Name: | solute carrier family 3 member 2 |
Gene Aliases: |
4F2, 4F2HC, 4T2HC, CD98, CD98HC, MDU1, NACAE |
Location: |
Chr.11:62856012-62888883 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC3A2 | NM_001012662.2 | 12 | 2041 | NP_001012680.1 |
| NM_001012664.2 | 10 | 1852 | NP_001012682.1 | |
| NM_001013251.2 | 9 | 1629 | NP_001013269.1 | |
| NM_002394.5 | 12 | 2038 | NP_002385.3 | |
| NR_037193.1 | 9 | 1371 | ||
| AA594112.1 | 1 | 312 | ||
| AB018010.1 | 9 | 1588 | BAA84649.1 | |
| AF055031.1 | 9 | 1341 | ||
| AK025584.1 | 12 | 1942 | ||
| AK090758.1 | 11 | 1202 | ||
| AK094620.1 | 6 | 1896 | ||
| AK225304.1 | 9 | 1576 | ||
| AK304497.1 | 10 | 1547 | ||
| BC001061.2 | 12 | 1948 | AAH01061.2 | |
| BC003000.1 | 10 | 1720 | AAH03000.2 | |
| BQ128156.1 | 1 | 316 | ||
| CA453972.1 | 4 | 561 | ||
| J02769.1 | 9 | 1530 | AAA51540.1 | |
| J02939.1 | 9 | 1586 | AAA52497.1 | |
| J03569.1 | 9 | 1615 | AAA35536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34011 | Chr.11:62885307 - 63226230 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map