Assay Details
Target Gene Details
Entrez Gene ID: | 54107 |
Gene Name: | DNA polymerase epsilon 3, accessory subunit |
Gene Aliases: |
CHARAC17, CHRAC17, YBL1, p17 |
Location: |
Chr.9:113407235-113410749 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLE3 | NM_001278255.1 | NP_001265184.1 | ||
| NM_017443.4 | NP_059139.3 | |||
| NR_027261.1 | ||||
| AF226077.1 | AAF72417.1 | |||
| AK074629.1 | BAC11099.1 | |||
| AK074762.1 | BAC11190.1 | |||
| AK074782.1 | BAC11206.1 | |||
| AK092840.1 | ||||
| AK096050.1 | ||||
| AY720898.1 | AAU15052.1 | |||
| BC003166.1 | AAH03166.1 | |||
| BC004170.2 | AAH04170.1 | |||
| DA477404.1 | ||||
| DA521953.1 | ||||
| DA908447.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621540 | Chr.9:113279513 - 113421563 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map