Assay Details
Target Gene Details
Entrez Gene ID: | 334 |
Gene Name: | amyloid beta precursor like protein 2 |
Gene Aliases: |
APLP-2, APPH, APPL2, CDEBP |
Location: |
Chr.11:130069821-130144811 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APLP2 | NM_001142276.1 | NP_001135748.1 | ||
| NM_001142277.1 | NP_001135749.1 | |||
| NM_001142278.1 | NP_001135750.1 | |||
| NM_001243299.1 | NP_001230228.1 | |||
| NM_001642.2 | NP_001633.1 | |||
| NR_024515.1 | ||||
| NR_024516.1 | ||||
| AF168956.1 | AAD47291.1 | |||
| AK056883.1 | ||||
| AK128162.1 | ||||
| AK225283.1 | ||||
| AK296765.1 | ||||
| AK300378.1 | ||||
| AK304736.1 | ||||
| AK308248.1 | ||||
| BC000373.2 | AAH00373.1 | |||
| BC004371.1 | AAH04371.1 | |||
| DC336210.1 | ||||
| L09209.1 | AAA35526.1 | |||
| L19597.1 | AAA35601.1 | |||
| L23114.1 | AAA36130.1 | |||
| L27631.1 | AAC41701.1 | |||
| S60099.1 | AAC60589.1 | |||
| Z22572.1 | CAA80295.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759871 | Chr.11:130099327 - 130472822 on Build GRCh38 | Loss |
|
| nsv832295 | Chr.11:130099212 - 130260193 on Build GRCh38 | Gain |
|
| nsv556565 | Chr.11:129996758 - 130180927 on Build GRCh38 | Loss |
|
| dgv251n67 | Chr.11:130005191 - 130124741 on Build GRCh38 | Gain |
|
| nsv549 | Chr.11:130104906 - 130137504 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map