Assay Details
Target Gene Details
Entrez Gene ID: | 84666 |
Gene Name: | resistin like beta |
Gene Aliases: |
FIZZ1, FIZZ2, HXCP2, RELM-beta, RELMb, RELMbeta, XCP2 |
Location: |
Chr.3:108755639-108757283 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RETNLB | NM_032579.2 | NP_115968.1 | ||
| AF290873.1 | AAK83105.1 | |||
| AF323084.1 | AAG59827.1 | |||
| AM050721.1 | CAJ19371.1 | |||
| AY358655.1 | AAQ89018.1 | |||
| BC069318.1 | AAH69318.1 | |||
| BC113502.1 | ||||
| BC113528.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2670614 | Chr.3:108747898 - 108755945 on Build GRCh38 | Deletion |
|
| nsv829659 | Chr.3:108654888 - 108818130 on Build GRCh38 | Gain |
|
| esv3568956 | Chr.3:108697937 - 108934542 on Build GRCh38 | Loss |
|
| nsv998974 | Chr.3:108720592 - 108813290 on Build GRCh38 | Gain |
|
| nsv829661 | Chr.3:108751158 - 108940686 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map