Assay Details
Target Gene Details
Entrez Gene ID: | 4717 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit C1 |
Gene Aliases: |
KFYI |
Location: |
Chr.4:139289917-139302551 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFC1 | NM_001184986.1 | 3 | 822 | NP_001171915.1 |
| NM_001184987.1 | 2 | 290 | NP_001171916.1 | |
| NM_001184988.1 | 3 | 326 | NP_001171917.1 | |
| NM_001184989.1 | 3 | 349 | NP_001171918.1 | |
| NM_001184990.1 | 2 | 286 | NP_001171919.1 | |
| NM_001184991.1 | 2 | 763 | NP_001171920.1 | |
| NM_002494.3 | 2 | 425 | NP_002485.1 | |
| AF047184.1 | AAC04269.1 | |||
| AF047435.1 | AAC39886.1 | |||
| AK291147.1 | ||||
| BC101798.1 | AAI01799.1 | |||
| BC107682.1 | 2 | 395 | AAI07683.1 | |
| BC112061.1 | AAI12062.1 | |||
| BG701467.1 | 2 | 430 | ||
| BM768825.1 | 3 | 326 | ||
| BU677336.1 | 4 | 398 | ||
| CA313899.1 | 4 | 435 | ||
| CB047240.1 | 4 | 445 | ||
| CB160001.1 | 3 | 277 | ||
| CN347371.1 | 2 | 191 | ||
| CR457005.1 | CAG33286.1 | |||
| CX163301.1 | 2 | 763 | ||
| W56292.1 |
More Information
Set Membership: |
|

Genomic Map