Assay Details
Target Gene Details
Entrez Gene ID: | 23359 |
Gene Name: | family with sequence similarity 189 member A1 |
Gene Aliases: |
TMEM228 |
Location: |
Chr.15:29120252-29570723 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM189A1 | NM_015307.1 | NP_056122.1 | ||
| XM_011521407.2 | XP_011519709.1 | |||
| DA188047.1 |
Target Gene Details
Entrez Gene ID: | 56160 |
Gene Name: | NSE3 homolog, SMC5-SMC6 complex component |
Gene Aliases: |
HCA4, MAGEG1, MAGEL3, NDNL2, NSE3 |
Location: |
Chr.15:29268149-29269816 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NSMCE3 | NM_138704.3 | 1 | 1107 | NP_619649.1 |
| AF490510.1 | 1 | 1115 | AAM08357.1 | |
| AK056957.1 | 1 | 1100 | BAB71325.1 | |
| AK074138.1 | 1 | 745 | BAB84964.1 | |
| BC041166.1 | 1 | 1063 | AAH41166.2 | |
| BC053999.1 | 1 | 1079 | AAH53999.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1040778 | Chr.15:28689598 - 30515727 on Build GRCh38 | Loss |
|
| nsv515660 | Chr.15:29250908 - 29272431 on Build GRCh38 | Gain |
|
| nsv527255 | Chr.15:28754022 - 30074044 on Build GRCh38 | Gain |
|
| nsv1039798 | Chr.15:28724126 - 30158154 on Build GRCh38 | Gain |
|
| nsv522597 | Chr.15:28724519 - 30074044 on Build GRCh38 | Loss |
|
| esv2761859 | Chr.15:29266382 - 29312594 on Build GRCh38 | Loss |
|
| nsv568675 | Chr.15:29035149 - 29916451 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115017847,rs115728036] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map