Assay Details
Target Gene Details
Entrez Gene ID: | 26127 |
Gene Name: | FGFR1 oncogene partner 2 |
Gene Aliases: |
HSPC123-like, WIT3.0 |
Location: |
Chr.12:26938372-26966648 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGFR1OP2 | NM_001171887.1 | 1 | 80 | NP_001165358.1 |
| NM_001171888.1 | 1 | 80 | NP_001165359.1 | |
| NM_015633.2 | 1 | 80 | NP_056448.1 | |
| AK001534.1 | 1 | 69 | BAA91745.1 | |
| DA388639.1 | 1 | 80 |
More Information
Set Membership: |
|

Genomic Map