Assay Details
Target Gene Details
Entrez Gene ID: | 81796 |
Gene Name: | solute carrier organic anion transporter family member 5A1 |
Gene Aliases: |
OATP-J, OATP-RP4, OATP5A1, OATPJ, OATPRP4, SLC21A15 |
Location: |
Chr.8:69667046-69835064 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLCO5A1 | NM_001146008.1 | 8 | 3382 | NP_001139480.1 |
| NM_001146009.1 | 8 | 3061 | NP_001139481.1 | |
| NM_030958.2 | 10 | 3437 | NP_112220.2 | |
| XM_005251313.2 | 9 | 3272 | XP_005251370.1 | |
| XM_017013883.1 | 9 | 3372 | XP_016869372.1 | |
| XM_017013885.1 | 9 | 3080 | XP_016869374.1 | |
| AF205075.1 | 10 | 3346 | AAG42207.1 | |
| AK022760.1 | 8 | 3382 | ||
| BC016067.2 | 1 | 640 | AAH16067.1 | |
| BC137424.1 | 9 | 3232 | ||
| BC139755.1 | ||||
| CA449383.1 | 1 | 370 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv92n68 | Chr.8:69559928 - 69745898 on Build GRCh38 | Loss |
|
| nsv6244 | Chr.8:69667061 - 69701867 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map