Assay Details
Target Gene Details
Entrez Gene ID: | 599 |
Gene Name: | BCL2 like 2 |
Gene Aliases: |
BCL-W, BCL2-L-2, BCLW, PPP1R51 |
Location: |
Chr.14:23306762-23311759 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL2L2 | NM_001199839.1 | 4 | 2522 | NP_001186768.1 |
| NM_004050.4 | 4 | 2567 | NP_004041.1 | |
| D87461.1 | 4 | 2514 |
Target Gene Details
Entrez Gene ID: | 100529063 |
Gene Name: | BCL2L2-PABPN1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.14:23306762-23326185 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL2L2-PABPN1 | NM_001199864.1 | NP_001186793.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs112933062] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map