Assay Details
Target Gene Details
Entrez Gene ID: | 29904 |
Gene Name: | eukaryotic elongation factor 2 kinase |
Gene Aliases: |
HSU93850, eEF-2K |
Location: |
Chr.16:22206264-22288745 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EEF2K | NM_013302.3 | 2 | 633 | NP_037434.1 |
| XM_017023193.1 | 2 | 344 | XP_016878682.1 | |
| XM_017023194.1 | 2 | 640 | XP_016878683.1 | |
| AK308757.1 | 2 | 578 | ||
| BC032665.1 | 3 | 344 | AAH32665.1 | |
| DA556486.1 | ||||
| DA735877.1 | 2 | 629 | ||
| U93850.1 | 1 | 148 | AAB58270.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469818 | Chr.16:22112125 - 22262126 on Build GRCh38 | Loss |
|
| esv3638195 | Chr.16:22082786 - 22273655 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map