Assay Details
Target Gene Details
Entrez Gene ID: | 8535 |
Gene Name: | chromobox 4 |
Gene Aliases: |
NBP16, PC2 |
Location: |
Chr.17:79833156-79839414 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CBX4 | NM_003655.2 | NP_003646.2 | ||
| XM_011525399.2 | XP_011523701.1 | |||
| AF013956.1 | AAB80718.1 | |||
| AY390430.1 | AAQ97596.1 | |||
| BC014967.1 | AAH14967.1 | |||
| BG029608.1 | ||||
| EU439707.1 | ||||
| KU178422.1 | ||||
| KU178423.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv538n67 | Chr.17:79765152 - 79848648 on Build GRCh38 | Gain |
|
| nsv469865 | Chr.17:79770348 - 79957778 on Build GRCh38 | Loss |
|
| nsv828120 | Chr.17:79833105 - 79842561 on Build GRCh38 | Gain |
|
| nsv828116 | Chr.17:79748992 - 79855333 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map