Assay Details
Target Gene Details
Entrez Gene ID: | 6782 |
Gene Name: | heat shock protein family A (Hsp70) member 13 |
Gene Aliases: |
STCH |
Location: |
Chr.21:14371115-14383188 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPA13 | NM_006948.4 | 5 | 3502 | NP_008879.3 |
| AI912654.1 | 1 | 488 | ||
| AK128540.1 | 5 | 3284 | ||
| BC036370.2 | 5 | 3464 | AAH36370.1 | |
| BE467288.1 | 1 | 187 |
More Information
Set Membership: |
|

Genomic Map