Assay Details
Target Gene Details
Entrez Gene ID: | 54495 |
Gene Name: | thioredoxin related transmembrane protein 3 |
Gene Aliases: |
PDIA13, TXNDC10 |
Location: |
Chr.18:68673688-68715116 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMX3 | NM_019022.3 | 16 | 2244 | NP_061895.3 |
| XM_005266714.2 | 15 | 2189 | XP_005266771.1 | |
| XM_011526033.2 | 14 | 2107 | XP_011524335.1 | |
| AB058733.1 | 16 | 2206 | ||
| AK000800.1 | 10 | 1684 | BAA91381.1 | |
| AK025025.1 | 1 | 137 | ||
| AK122715.1 | 16 | 2236 | ||
| AL832341.1 | 16 | 2192 | ||
| AL832356.1 | 16 | 2192 | ||
| BX647846.1 | 16 | 2253 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893127 | Chr.18:68641143 - 68863126 on Build GRCh38 | Gain |
|
| dgv3390n100 | Chr.18:68019389 - 68866022 on Build GRCh38 | Gain |
|
| esv2758736 | Chr.18:68308501 - 68725725 on Build GRCh38 | Gain+Loss |
|
| nsv458117 | Chr.18:68025058 - 68867575 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map