Assay Details
Target Gene Details
Entrez Gene ID: | 54058 |
Gene Name: | chromosome 21 open reading frame 58 |
Gene Aliases: |
- |
Location: |
Chr.21:46300426-46323899 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C21orf58 | NM_001286462.1 | NP_001273391.1 | ||
| NM_001286463.1 | 7 | 1245 | NP_001273392.1 | |
| NM_001286476.1 | NP_001273405.1 | |||
| NM_001286477.1 | 8 | 1886 | NP_001273406.1 | |
| NM_058180.4 | 8 | 2258 | NP_478060.2 | |
| XM_005261148.4 | XP_005261205.1 | |||
| XM_006724018.3 | XP_006724081.1 | |||
| XM_011529619.2 | XP_011527921.1 | |||
| XM_011529620.2 | XP_011527922.1 | |||
| XM_011529621.2 | 8 | 2241 | XP_011527923.1 | |
| XM_011529623.2 | 8 | 4246 | XP_011527925.1 | |
| XM_011529624.2 | 8 | 1121 | XP_011527926.1 | |
| XM_011529625.2 | 8 | 1395 | XP_011527927.1 | |
| XM_011529626.2 | 9 | 4592 | XP_011527928.1 | |
| XM_011529627.2 | 9 | 4672 | XP_011527929.1 | |
| XM_017028388.1 | 9 | 4423 | XP_016883877.1 | |
| XM_017028389.1 | XP_016883878.1 | |||
| XM_017028390.1 | 9 | 2590 | XP_016883879.1 | |
| XM_017028391.1 | 8 | 2413 | XP_016883880.1 | |
| XM_017028392.1 | 10 | 1610 | XP_016883881.1 | |
| XM_017028393.1 | 9 | 1409 | XP_016883882.1 | |
| AK056387.1 | 8 | 2231 | ||
| AK096641.1 | 7 | 2162 | ||
| AK098098.1 | 7 | 1182 | BAC05232.1 | |
| AY039243.1 | AAK72408.1 | |||
| AY039244.1 | AAK72409.1 | |||
| BM475055.1 | ||||
| CB305953.1 |
Target Gene Details
Entrez Gene ID: | 54059 |
Gene Name: | ybeY metallopeptidase (putative) |
Gene Aliases: |
C21orf57 |
Location: |
Chr.21:46286056-46314188 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YBEY | XM_011529633.2 | XP_011527935.1 | ||
| XM_017028394.1 | XP_016883883.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519904 | Chr.21:46163592 - 46461396 on Build GRCh38 | Loss |
|
| nsv3547 | Chr.21:46283787 - 46321516 on Build GRCh38 | Insertion |
|
| nsv587980 | Chr.21:46295382 - 46302616 on Build GRCh38 | Loss |
|
| nsv527355 | Chr.21:46258860 - 46443112 on Build GRCh38 | Gain |
|
| nsv470910 | Chr.21:45933861 - 46665076 on Build GRCh38 | Gain |
|
| nsv587975 | Chr.21:46241321 - 46410929 on Build GRCh38 | Gain |
|
| nsv1060978 | Chr.21:46068159 - 46413726 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map