Assay Details
Target Gene Details
Entrez Gene ID: | 26580 |
Gene Name: | BSCL2, seipin lipid droplet biogenesis associated |
Gene Aliases: |
GNG3LG, HMN5, PELD, SPG17 |
Location: |
Chr.11:62690262-62709619 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BSCL2 | NM_001122955.3 | 7 | 1444 | NP_001116427.1 |
| NM_001130702.2 | NP_001124174.2 | |||
| NM_032667.6 | 7 | 958 | NP_116056.3 | |
| NR_037948.1 | 8 | 1534 | ||
| NR_037949.1 | 8 | 1534 | ||
| AF052149.1 | 7 | 1084 | ||
| AK027524.1 | BAB55175.1 | |||
| AK075317.1 | 7 | 941 | BAC11543.1 | |
| AK223606.1 | 7 | 1135 | BAD97326.1 | |
| AK225029.1 | 7 | 939 | ||
| AK315423.1 | 8 | 1451 | ||
| BC004911.2 | 7 | 937 | AAH04911.1 | |
| BC012140.1 | 7 | 928 | AAH12140.1 | |
| BC041640.1 | 7 | 910 | AAH41640.1 | |
| BC093048.1 | 7 | 993 | AAH93048.1 | |
| KU178595.1 | 6 | 740 | ||
| KU178596.1 | 5 | 658 |
Target Gene Details
Entrez Gene ID: | 100534595 |
Gene Name: | HNRNPUL2-BSCL2 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.11:62690262-62727384 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPUL2-BSCL2 | NR_037946.1 | 20 | 3452 | |
| AK122942.1 | 20 | 3452 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832182 | Chr.11:62528103 - 62695376 on Build GRCh38 | Gain+Loss |
|
| nsv555174 | Chr.11:62566624 - 62729047 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map