Assay Details
Target Gene Details
Entrez Gene ID: | 55661 |
Gene Name: | DEAD-box helicase 27 |
Gene Aliases: |
DRS1, Drs1p, HSPC259, PP3241, RHLP, dJ686N3.1 |
Location: |
Chr.20:49219295-49244077 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX27 | NM_017895.7 | NP_060365.7 | ||
| XM_011528888.2 | XP_011527190.1 | |||
| AK022979.1 | BAB14343.1 | |||
| AK300135.1 | ||||
| AL832131.1 | ||||
| AY044431.1 | AAK95821.1 | |||
| BC009304.1 | AAH09304.2 | |||
| BC011927.2 | AAH11927.2 | |||
| BC016060.1 | AAH16060.2 | |||
| BC104480.1 | AAI04481.1 | |||
| BC126287.1 | ||||
| BC130275.1 | ||||
| BC144125.1 | ||||
| BF026095.1 | ||||
| DB442175.2 | ||||
| EU718675.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064443 | Chr.20:49135574 - 49439352 on Build GRCh38 | Gain |
|
| nsv1067146 | Chr.20:48878894 - 49475527 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map