Assay Details
Target Gene Details
Entrez Gene ID: | 27035 |
Gene Name: | NADPH oxidase 1 |
Gene Aliases: |
GP91-2, MOX1, NOH-1, NOH1 |
Location: |
Chr.X:100843324-100874345 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOX1 | NM_001271815.1 | NP_001258744.1 | ||
| NM_007052.4 | NP_008983.2 | |||
| NM_013955.2 | NP_039249.1 | |||
| XM_017029407.1 | XP_016884896.1 | |||
| AF166326.1 | AAF23232.1 | |||
| AF166327.1 | AAF23233.1 | |||
| AF166328.1 | AAF23234.1 | |||
| AJ438989.1 | CAD27735.1 | |||
| AK292201.1 | ||||
| AK313678.1 | ||||
| BC075014.2 | AAH75014.1 | |||
| BC075015.2 | AAH75015.1 | |||
| BX956931.1 | ||||
| DQ424904.1 | ABD83669.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7011 | Chr.X:100857971 - 100890941 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map