Assay Details
Target Gene Details
Entrez Gene ID: | 283899 |
Gene Name: | INO80 complex subunit E |
Gene Aliases: |
CCDC95 |
Location: |
Chr.16:29996209-30005794 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INO80E | NM_001304562.1 | NP_001291491.1 | ||
| NM_001304563.1 | NP_001291492.1 | |||
| NM_173618.2 | NP_775889.1 | |||
| NR_130786.1 | ||||
| NR_130787.1 | ||||
| NR_134855.1 | ||||
| XM_011545809.2 | XP_011544111.1 | |||
| XM_011545811.2 | XP_011544113.1 | |||
| XM_011545812.2 | XP_011544114.1 | |||
| XM_017023169.1 | XP_016878658.1 | |||
| AK074137.1 | BAB84963.1 | |||
| AK075133.1 | BAC11424.1 | |||
| AK130409.1 | ||||
| AK309498.1 | ||||
| AL525020.3 | ||||
| AY189289.1 | AAO86733.1 | |||
| BC047712.2 | AAH47712.1 | |||
| BI093312.1 | ||||
| BU528528.1 | ||||
| BX356644.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3638378 | Chr.16:29973809 - 30006402 on Build GRCh38 | Loss |
|
| esv997840 | Chr.16:29991169 - 30003917 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map