Assay Details
Target Gene Details
Entrez Gene ID: | 84109 |
Gene Name: | pyroglutamylated RFamide peptide receptor |
Gene Aliases: |
AQ27, GPR103, SP9155 |
Location: |
Chr.4:121328642-121381029 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QRFPR | NM_198179.2 | NP_937822.2 | ||
| XM_017008693.1 | XP_016864182.1 | |||
| AB109629.1 | BAC98938.1 | |||
| AK309337.1 | 2 | 895 | ||
| BC128133.1 | ||||
| BG169612.1 | ||||
| JF810892.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4494 | Chr.4:121335683 - 121369306 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map