Assay Details
Target Gene Details
Entrez Gene ID: | 64645 |
Gene Name: | major facilitator superfamily domain containing 14A |
Gene Aliases: |
HIAT1 |
Location: |
Chr.1:100038078-100083377 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD14A | NM_033055.2 | 12 | 2096 | NP_149044.2 |
| XM_005271118.1 | 13 | 2372 | XP_005271175.1 | |
| XM_011541957.1 | 11 | 2137 | XP_011540259.1 | |
| XM_017002084.1 | 13 | 2339 | XP_016857573.1 | |
| AK075208.1 | 6 | 1297 | BAC11473.1 | |
| AK096669.1 | 9 | 1747 | BAC04836.1 | |
| AK127671.1 | 1 | 1608 | ||
| AL137576.1 | 4 | 1190 | CAB70819.2 | |
| BC064409.1 | 9 | 1753 | AAH64409.1 | |
| BC107768.1 | 9 | 1845 | AAI07769.1 |
Target Gene Details
Entrez Gene ID: | 163786 |
Gene Name: | SAS-6 centriolar assembly protein |
Gene Aliases: |
MCPH14, SAS-6, SAS6 |
Location: |
Chr.1:100082632-100133095 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SASS6 | XM_017000486.1 | 17 | 4517 | XP_016855975.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830804 | Chr.1:100036777 - 100210481 on Build GRCh38 | Gain |
|
| nsv546888 | Chr.1:99954311 - 100084889 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map