Assay Details
Target Gene Details
Entrez Gene ID: | 4601 |
Gene Name: | MAX interactor 1, dimerization protein |
Gene Aliases: |
MAD2, MXD2, MXI, bHLHc11 |
Location: |
Chr.10:110207605-110287365 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MXI1 | NM_130439.3 | 1 | 398 | NP_569157.2 |
| BC035128.1 | 1 | 404 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2673748 | Chr.10:110207787 - 110211534 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map