Assay Details
Target Gene Details
Entrez Gene ID: | 7071 |
Gene Name: | Kruppel like factor 10 |
Gene Aliases: |
EGR-alpha, EGRA, TIEG, TIEG1 |
Location: |
Chr.8:102648777-102655902 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLF10 | NM_001032282.3 | 4 | 2291 | NP_001027453.1 |
| NM_005655.3 | 4 | 2133 | NP_005646.1 | |
| NR_103759.1 | 3 | 1378 | ||
| NR_103760.1 | 4 | 1501 | ||
| BC011538.1 | 4 | 2118 | AAH11538.1 | |
| BC095399.1 | 4 | 1954 | AAH95399.1 | |
| S81439.1 | 4 | 2267 | AAB36088.1 | |
| U21847.1 | 4 | 1918 | AAC50340.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33118 | Chr.8:102346339 - 103055944 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map