Assay Details
Target Gene Details
Entrez Gene ID: | 55291 |
Gene Name: | protein phosphatase 6 regulatory subunit 3 |
Gene Aliases: |
C11orf23, PP6R3, SAP190, SAPL, SAPLa, SAPS3 |
Location: |
Chr.11:68460712-68615334 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2744662 | Chr.11:68515367 - 68644348 on Build GRCh38 | Deletion |
|
| nsv832195 | Chr.11:68518915 - 68707143 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map