Assay Details
Target Gene Details
Entrez Gene ID: | 84658 |
Gene Name: | adhesion G protein-coupled receptor E3 |
Gene Aliases: |
EMR3 |
Location: |
Chr.19:14600175-14674918 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRE3 | AL832663.1 |
Target Gene Details
Entrez Gene ID: | 388512 |
Gene Name: | C-type lectin domain family 17 member A |
Gene Aliases: |
- |
Location: |
Chr.19:14580777-14612035 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC17A | NM_001204118.1 | 14 | 1354 | NP_001191047.1 |
| NM_207390.3 | 13 | 1244 | NP_997273.3 | |
| NR_109784.1 | 14 | 1417 | ||
| NR_109785.1 | 13 | 1202 | ||
| XM_017026785.1 | 15 | 1898 | XP_016882274.1 | |
| XM_017026786.1 | 15 | 1762 | XP_016882275.1 | |
| XM_017026787.1 | 16 | 2117 | XP_016882276.1 | |
| XM_017026788.1 | 15 | 1874 | XP_016882277.1 | |
| XM_017026789.1 | 11 | 1576 | XP_016882278.1 | |
| XM_017026790.1 | 14 | 1850 | XP_016882279.1 | |
| XM_017026791.1 | 13 | 1699 | XP_016882280.1 | |
| BC140848.1 | 14 | 1315 | ||
| BC144665.1 | 13 | 1163 |
More Information
Set Membership: |
|

Genomic Map