Assay Details
Target Gene Details
Entrez Gene ID: | 10873 |
Gene Name: | malic enzyme 3 |
Gene Aliases: |
NADP-ME |
Location: |
Chr.11:86441108-86674745 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ME3 | NM_001014811.1 | 14 | 2049 | NP_001014811.1 |
| NM_001161586.1 | 15 | 2122 | NP_001155058.1 | |
| NM_006680.2 | 15 | 1897 | NP_006671.2 | |
| XM_005273717.1 | 15 | 1998 | XP_005273774.1 | |
| XM_017017136.1 | 15 | 2880 | XP_016872625.1 | |
| XM_017017137.1 | 11 | 1422 | XP_016872626.1 | |
| XM_017017138.1 | 16 | 2006 | XP_016872627.1 | |
| AI560417.1 | 1 | 190 | ||
| AI948779.1 | 1 | 192 | ||
| AK313695.1 | ||||
| BC022472.2 | 14 | 2049 | AAH22472.1 | |
| BQ882896.1 | 6 | 782 | ||
| CB306059.1 | 1 | 208 | ||
| X79440.1 | 14 | 1827 | CAA55956.1 |
More Information
Set Membership: |
|

Genomic Map