Assay Details
Target Gene Details
Entrez Gene ID: | 10488 |
Gene Name: | cAMP responsive element binding protein 3 |
Gene Aliases: |
LUMAN, LZIP, sLZIP |
Location: |
Chr.9:35732320-35737008 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREB3 | NM_006368.4 | 9 | 1794 | NP_006359.3 |
| AF009368.1 | 9 | 1354 | AAB69652.1 | |
| AF211848.1 | 9 | 1779 | AAG43528.1 | |
| BQ014045.1 | 1 | 75 |
Target Gene Details
Entrez Gene ID: | 57704 |
Gene Name: | glucosylceramidase beta 2 |
Gene Aliases: |
AD035, NLGase, SPG46 |
Location: |
Chr.9:35736862-35749255 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GBA2 | NM_020944.2 | 17 | 3526 | NP_065995.1 |
| XM_005251526.4 | 17 | 3505 | XP_005251583.1 | |
| XM_006716809.3 | 17 | 3571 | XP_006716872.1 | |
| XM_011517973.2 | 17 | 3288 | XP_011516275.1 | |
| XM_017014937.1 | 17 | 3487 | XP_016870426.1 | |
| XM_017014938.1 | 17 | 3728 | XP_016870427.1 | |
| XM_017014939.1 | 17 | 3222 | XP_016870428.1 | |
| XM_017014940.1 | 16 | 2912 | XP_016870429.1 | |
| XM_017014942.1 | 17 | 3144 | XP_016870431.1 | |
| XM_017014943.1 | 17 | 3126 | XP_016870432.1 | |
| XM_017014944.1 | 16 | 2589 | XP_016870433.1 | |
| XM_017014945.1 | 16 | 2571 | XP_016870434.1 | |
| XM_017014946.1 | 18 | 3182 | XP_016870435.1 | |
| AB046825.1 | 17 | 3673 | BAB13431.1 | |
| AJ309567.1 | 17 | 3526 | CAC83792.1 | |
| AK027884.1 | 17 | 3189 | BAB55430.1 | |
| AK057610.1 | 2 | 1588 | ||
| AK295967.1 | ||||
| AK297435.1 | 15 | 2533 | ||
| AL834306.1 | 17 | 3286 | CAD38976.1 | |
| BC011363.1 | 17 | 3485 | AAH11363.1 |
More Information
Set Membership: |
|

Genomic Map