Assay Details
Target Gene Details
Entrez Gene ID: | 3242 |
Gene Name: | 4-hydroxyphenylpyruvate dioxygenase |
Gene Aliases: |
4-HPPD, 4HPPD, GLOD3, HPPDASE, PPD |
Location: |
Chr.12:121839527-121888611 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPD | NM_001171993.1 | NP_001165464.1 | ||
| NM_002150.2 | NP_002141.1 | |||
| AK057510.1 | ||||
| AK290826.1 | ||||
| BC024287.1 | AAH24287.1 | |||
| X72389.1 | CAA51082.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160065 | Chr.12:121780583 - 121859371 on Build GRCh38 | Deletion |
|
| esv3892268 | Chr.12:121747658 - 121845875 on Build GRCh38 | Loss |
|
| nsv912 | Chr.12:121841785 - 121873999 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map