Assay Details
Target Gene Details
Entrez Gene ID: | 26073 |
Gene Name: | DNA polymerase delta interacting protein 2 |
Gene Aliases: |
PDIP38, POLD4, p38 |
Location: |
Chr.17:28346628-28357589 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLDIP2 | NM_001290145.1 | NP_001277074.1 | ||
| NM_015584.4 | NP_056399.1 | |||
| AF077203.1 | AAD26998.1 | |||
| AF179891.1 | AAK92018.1 | |||
| AK293707.1 | ||||
| AK313232.1 | ||||
| BC000655.2 | AAH00655.1 | |||
| BC009265.2 | AAH09265.1 | |||
| BU932070.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2017 | Chr.17:28325611 - 28357716 on Build GRCh38 | Insertion |
|
| nsv1056839 | Chr.17:28348199 - 28428188 on Build GRCh38 | Gain |
|
| nsv519388 | Chr.17:28303453 - 28379927 on Build GRCh38 | Loss |
|
| nsv574653 | Chr.17:28227749 - 28501479 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map