Assay Details
Target Gene Details
Entrez Gene ID: | 101927718 |
Gene Name: | DSG1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.18:31343368-31426988 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSG1-AS1 | NR_110788.1 | |||
| BX476374.1 | ||||
| BX476414.1 |
Target Gene Details
Entrez Gene ID: | 147409 |
Gene Name: | desmoglein 4 |
Gene Aliases: |
CDGF13, CDHF13, HYPT6, LAH |
Location: |
Chr.18:31376777-31415791 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSG4 | NM_001134453.2 | 1 | 84 | NP_001127925.1 |
| NM_177986.4 | 1 | 84 | NP_817123.1 | |
| AY168788.1 | AAO43657.1 | |||
| AY177664.1 | 1 | 84 | AAO47078.1 | |
| AY228236.1 | AAP49811.1 | |||
| BC132907.1 | ||||
| BC132909.1 |
More Information
Set Membership: |
|

Genomic Map