Assay Details
Target Gene Details
Entrez Gene ID: | 5985 |
Gene Name: | replication factor C subunit 5 |
Gene Aliases: |
RFC36 |
Location: |
Chr.12:118016701-118033568 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFC5 | NM_001130112.2 | NP_001123584.1 | ||
| NM_001206801.1 | NP_001193730.1 | |||
| NM_007370.5 | NP_031396.1 | |||
| NM_181578.3 | NP_853556.2 | |||
| XM_011538643.2 | XP_011536945.1 | |||
| XM_011538645.2 | XP_011536947.1 | |||
| XM_017019779.1 | XP_016875268.1 | |||
| AB208992.1 | BAD92229.1 | |||
| AK094575.1 | ||||
| AK290685.1 | ||||
| AK291107.1 | ||||
| AU121576.1 | ||||
| BC001866.1 | AAH01866.1 | |||
| BC013961.2 | AAH13961.1 | |||
| CR407651.1 | CAG28579.1 | |||
| DB270923.1 | ||||
| L07540.1 | AAB09784.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581916 | Chr.12:117356518 - 118228514 on Build GRCh38 | Gain |
|
| nsv560388 | Chr.12:117953795 - 118044547 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map