Assay Details
Target Gene Details
Entrez Gene ID: | 51621 |
Gene Name: | Kruppel like factor 13 |
Gene Aliases: |
BTEB3, FKLF2, NSLP1, RFLAT-1, RFLAT1 |
Location: |
Chr.15:31326855-31435665 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLF13 | NM_001302461.1 | 1 | 392 | NP_001289390.1 |
| NM_015995.3 | 1 | 392 | NP_057079.2 | |
| AF132599.1 | 1 | 406 | AAD26864.1 | |
| AF150628.1 | 1 | 80 | AAD34020.1 | |
| BC010438.1 | 1 | 347 | AAH10438.1 | |
| BC012741.2 | 1 | 250 | AAH12741.1 | |
| BC013946.2 | 1 | 347 | AAH13946.1 | |
| KJ534876.1 | ||||
| KJ535015.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636052 | Chr.15:31325578 - 31378949 on Build GRCh38 | Gain |
|
| nsv1070262 | Chr.15:31326896 - 31328097 on Build GRCh38 | Deletion |
|
| dgv4434n54 | Chr.15:31326533 - 31328530 on Build GRCh38 | Loss |
|
| esv29856 | Chr.15:31325628 - 31328835 on Build GRCh38 | Loss |
|
| nsv568799 | Chr.15:31326697 - 31328582 on Build GRCh38 | Gain |
|
| nsv827280 | Chr.15:31325333 - 31329744 on Build GRCh38 | Gain |
|
| dgv4429n54 | Chr.15:31325585 - 31327742 on Build GRCh38 | Loss |
|
| nsv516681 | Chr.15:31324471 - 31383250 on Build GRCh38 | Loss |
|
| esv34100 | Chr.15:31204311 - 31500388 on Build GRCh38 | Loss |
|
| dgv4433n54 | Chr.15:31325910 - 31328083 on Build GRCh38 | Gain |
|
| nsv456747 | Chr.15:31245293 - 31345793 on Build GRCh38 | Loss |
|
| dgv4432n54 | Chr.15:31325674 - 31329118 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map