Assay Details
Target Gene Details
Entrez Gene ID: | 27330 |
Gene Name: | ribosomal protein S6 kinase A6 |
Gene Aliases: |
PP90RSK4, RSK4 |
Location: |
Chr.X:84058346-84187935 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 24 - Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS6KA6 | NM_014496.4 | NP_055311.1 | ||
| XM_011530917.2 | XP_011529219.1 | |||
| XM_011530918.2 | XP_011529220.1 | |||
| XM_011530919.2 | XP_011529221.1 | |||
| XM_011530920.2 | XP_011529222.1 | |||
| XM_017029423.1 | XP_016884912.1 | |||
| XM_017029424.1 | XP_016884913.1 | |||
| XM_017029425.1 | XP_016884914.1 | |||
| XM_017029426.1 | XP_016884915.1 | |||
| AF184965.1 | AAF13190.1 | |||
| AK313240.1 | ||||
| BC143648.1 | ||||
| CR536566.1 | CAG38803.1 |
More Information
Set Membership: |
|

Genomic Map