Assay Details
Target Gene Details
Entrez Gene ID: | 1536 |
Gene Name: | cytochrome b-245 beta chain |
Gene Aliases: |
AMCBX2, CGD, GP91-1, GP91-PHOX, GP91PHOX, IMD34, NOX2, p91-PHOX |
Location: |
Chr.X:37780017-37813461 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYBB | NM_000397.3 | NP_000388.2 | ||
| AK289753.1 | ||||
| AK301476.1 | ||||
| AK301491.1 | ||||
| AK304033.1 | ||||
| AK312929.1 | ||||
| BC032720.1 | AAH32720.1 | |||
| DA995835.1 | ||||
| KU178009.1 | ||||
| KU178010.1 | ||||
| X04011.1 | CAA27635.1 |
More Information
Set Membership: |
|

Genomic Map