Assay Details
Target Gene Details
Entrez Gene ID: | 50937 |
Gene Name: | cell adhesion associated, oncogene regulated |
Gene Aliases: |
CDO, CDON1, HPE11, ORCAM |
Location: |
Chr.11:125956813-126063335 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDON | NM_001243597.1 | 20 | 6970 | NP_001230526.1 |
| NM_016952.4 | 20 | 6901 | NP_058648.4 | |
| XM_011542862.2 | 20 | 7131 | XP_011541164.1 | |
| XM_011542863.2 | 20 | 7135 | XP_011541165.1 | |
| XM_011542864.2 | 20 | 7048 | XP_011541166.1 | |
| XM_011542865.2 | 20 | 6931 | XP_011541167.1 | |
| XM_011542866.2 | 20 | 7062 | XP_011541168.1 | |
| XM_017017873.1 | 20 | 6967 | XP_016873362.1 | |
| AB209396.1 | 11 | 5290 | BAD92633.1 | |
| AK022986.1 | 1 | 2143 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv535 | Chr.11:125918491 - 125963371 on Build GRCh38 | Deletion |
|
| nsv556525 | Chr.11:125690285 - 125971104 on Build GRCh38 | Gain |
|
| esv32964 | Chr.11:125495069 - 126046387 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map