Assay Details
Target Gene Details
Entrez Gene ID: | 283600 |
Gene Name: | solute carrier family 25 member 47 |
Gene Aliases: |
C14orf68, HDMCP, HMFN1655 |
Location: |
Chr.14:100323342-100330378 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A47 | NM_207117.2 | 6 | 1119 | NP_997000.2 |
| AB073385.1 | 7 | 1043 | BAD38636.1 | |
| AK092001.1 | 6 | 1123 | ||
| AY569438.1 | 6 | 1119 | AAS80155.1 | |
| BC137253.1 | 6 | 1099 | ||
| BC137254.1 | 6 | 1099 | ||
| BX248260.1 | 6 | 1108 | CAD62588.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832875 | Chr.14:100192592 - 100368905 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map