Assay Details
Target Gene Details
Entrez Gene ID: | 6520 |
Gene Name: | solute carrier family 3 member 2 |
Gene Aliases: |
4F2, 4F2HC, 4T2HC, CD98, CD98HC, MDU1, NACAE |
Location: |
Chr.11:62856012-62888883 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC3A2 | NM_001012662.2 | NP_001012680.1 | ||
| NM_001012664.2 | NP_001012682.1 | |||
| NM_001013251.2 | NP_001013269.1 | |||
| NM_002394.5 | NP_002385.3 | |||
| NR_037193.1 | ||||
| AB018010.1 | BAA84649.1 | |||
| AF055031.1 | ||||
| AK025584.1 | ||||
| AK090758.1 | ||||
| AK094620.1 | ||||
| AK225304.1 | ||||
| AK304497.1 | ||||
| AK308797.1 | ||||
| AK311592.1 | ||||
| BC001061.2 | AAH01061.2 | |||
| BC003000.1 | AAH03000.2 | |||
| CA453972.1 | ||||
| J02769.1 | AAA51540.1 | |||
| J02939.1 | AAA52497.1 | |||
| J03569.1 | AAA35536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34011 | Chr.11:62885307 - 63226230 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map