Assay Details
Target Gene Details
Entrez Gene ID: | 4695 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A2 |
Gene Aliases: |
B8, CD14, CIB8 |
Location: |
Chr.5:140645363-140647785 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFA2 | NM_001185012.1 | 1 | 186 | NP_001171941.1 |
| NM_002488.4 | 1 | 186 | NP_002479.1 | |
| NR_033697.1 | 1 | 186 | ||
| AF047185.1 | AAC04270.1 | |||
| AF077029.1 | 1 | 57 | AAD27762.1 | |
| BC003674.1 | AAH03674.1 | |||
| BM706616.1 | 1 | 180 | ||
| BQ054127.1 | ||||
| CR457016.1 | CAG33297.1 |
More Information
Set Membership: |
|

Genomic Map