Assay Details
Target Gene Details
Entrez Gene ID: | 143 |
Gene Name: | poly(ADP-ribose) polymerase family member 4 |
Gene Aliases: |
ADPRTL1, ARTD4, PARP-4, PARPL, PH5P, VAULT3, VPARP, VWA5C, p193 |
Location: |
Chr.13:24420930-24512993 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARP4 | NM_006437.3 | 8 | 936 | NP_006428.2 |
| XM_011534931.1 | 7 | 986 | XP_011533233.1 | |
| XM_011534932.2 | 8 | 1153 | XP_011533234.1 | |
| AF057160.1 | 8 | 882 | AAC62491.1 | |
| AF158255.1 | 8 | 936 | AAD47250.1 | |
| AK304399.1 | 8 | 904 | ||
| BC017815.1 | 8 | 875 | ||
| D79999.2 | 7 | 835 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892310 | Chr.13:24212887 - 24577193 on Build GRCh38 | Gain |
|
| nsv832567 | Chr.13:24302981 - 24520733 on Build GRCh38 | Loss |
|
| esv2761200 | Chr.13:24313097 - 24636778 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map